missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(3,4-Dichlorophenyl)carbazate 98.0+%, TCI America™

Catalog No. E09981G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09981G 1 g
E09985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09981G Supplier TCI America Supplier No. E09981G

Chemical Identifiers

CAS 13124-15-7
Molecular Formula C9H10Cl2N2O2
Molecular Weight (g/mol) 249.09
MDL Number MFCD28975114
InChI Key JLVAIPOSBJQZGC-UHFFFAOYSA-N
Synonym 3-(3,4-Dichlorophenyl)carbazic Acid Ethyl Ester, 1-(3,4-Dichlorophenyl)-2-(ethoxycarbonyl)hydrazine
PubChem CID 12787057
IUPAC Name N'-(3,4-dichlorophenyl)ethoxycarbohydrazide
SMILES CCOC(=O)NNC1=CC(Cl)=C(Cl)C=C1

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 1 g
Formula Weight 249.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-(3,4-Dichlorophenyl)carbazate