missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(2-Pyridylamino)propionate 98.0+%, TCI America™

Catalog No. E12145G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12145G 5 g
E121425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12145G Supplier TCI America Supplier No. E12145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103041-38-9 | C10H14N2O2 | 194.234 g/mol

Chemical Identifiers

CAS 103041-38-9
Molecular Formula C10H14N2O2
Molecular Weight (g/mol) 194.234
MDL Number MFCD09833621
InChI Key UITNIDFEANEWPC-UHFFFAOYSA-N
Synonym 3-(2-Pyridylamino)propionic Acid Ethyl Ester
PubChem CID 11984503
IUPAC Name ethyl 3-(pyridin-2-ylamino)propanoate
SMILES CCOC(=O)CCNC1=CC=CC=N1

Specifications

Melting Point 53°C
Boiling Point 125°C
Color White-Yellow
Quantity 5 g
Formula Weight 194.23
Percent Purity ≥98.0% (T)
Physical Form Crystal/Lump
Chemical Name or Material Ethyl 3-(2-Pyridylamino)propionate