CAS RN 103041-38-9
IUPAC Name:
ethyl 3-[(pyridin-2-yl)amino]propanoate
Synonyms:
ethyl 3-(pyridin-2-ylamino)propanoate
Molecular Weight (g/mol):
194.23
Molecular Formula:
C10H14N2O2
InChi Key:
UITNIDFEANEWPC-UHFFFAOYSA-N
SMILES:
CCOC(=O)CCNC1=CC=CC=N1
1
–
1
of
1
results
1
Ethyl 3-(2-Pyridylamino)propionate 98.0+%, TCI America™
CAS: 103041-38-9 Molecular Formula: C10H14N2O2 Molecular Weight (g/mol): 194.234 MDL Number: MFCD09833621 InChI Key: UITNIDFEANEWPC-UHFFFAOYSA-N Synonym: 3-(2-Pyridylamino)propionic Acid Ethyl Ester PubChem CID: 11984503 IUPAC Name: ethyl 3-(pyridin-2-ylamino)propanoate SMILES: CCOC(=O)CCNC1=CC=CC=N1