missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(2-bromophenyl)propionate, 98%, Thermo Scientific Chemicals

Catalog No. p-7055028
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6350506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6350506 Supplier Thermo Scientific Chemicals Supplier No. H6350506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 135613-33-1 | Molecular Formula: C11H13BrO2 | Molecular Weight: 257.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 135613-33-1
Molecular Formula C11H13BrO2
Molecular Weight (g/mol) 257.127
MDL Number MFCD07772962
InChI Key ZKICRDRBPPOQLM-UHFFFAOYSA-N
Synonym ethyl 3-2-bromophenyl propanoate, 3-2-bromo-phenyl-propionic acid ethyl ester, 3-2-bromophenyl propionic acid ethyl ester, benzenepropanoic acid, 2-bromo-, ethyl ester, ksc497c5h, ethyl3-2-bromophenyl propanoate, ethyl 2-bromo-3-benzenepropanoate, 2-bromo-hydrocinnamic acid ethyl ester, 3-2-bromophenyl propanoic acid ethyl ester
PubChem CID 10611137
IUPAC Name ethyl 3-(2-bromophenyl)propanoate
SMILES CCOC(=O)CCC1=CC=CC=C1Br

Specifications

Color Yellow
Quantity 5 g
Formula Weight 257.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 3-(2-bromophenyl)propionate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only