missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-(2,6-Dichloro-5-fluoro-3-pyridyl)-3-oxopropionate 98.0+%, TCI America™
Supplier: TCI America E081125G
Specifications
| Ethyl 3-(2,6-Dichloro-5-fluoro-3-pyridyl)-3-oxopropionate | |
| 69°C | |
| MFCD00799535 | |
| ethyl 3-2,6-dichloro-5-fluoropyridin-3-yl-3-oxopropanoate, ethyl 3-2,6-dichloro-5-fluoro-3-pyridyl-3-oxopropionate, ethyl 2,6-dichloro-5-fluoronicotinoylacetate, ethyl 2,6-dichloro-5-fluoro pyridine-3-acetoacetate, ethyl 2,6-dichloro-5-fluoronicotinoyl acetate, ethyl 2,6-dichloro-5-fluornicotinoylacetate, ethyl 3-2,6-dichloro-5-fluoro-3-pyridiyl-3-oxopropanoate, 2,6-dichloro-5-fluoronicotinoyl acetic acid ethyl ester, ethyl 2,6-dichloro-5-fluoro nicotinylacetate, ethyl 3-2,6-dichloro-5-fluoro 3-pyridyl-3-oxopropanoate | |
| CCOC(=O)CC(=O)C1=CC(F)=C(Cl)N=C1Cl | |
| 280.08 | |
| 280.08 | |
| Crystalline Powder |
| 96568-04-6 | |
| C10H8Cl2FNO3 | |
| 25 g | |
| IEUHWNLWVMLHHC-UHFFFAOYSA-N | |
| ethyl 3-(2,6-dichloro-5-fluoropyridin-3-yl)-3-oxopropanoate | |
| 2733708 | |
| ≥98.0% (T) |
Chemical Identifiers
| 96568-04-6 | |
| 280.08 | |
| IEUHWNLWVMLHHC-UHFFFAOYSA-N | |
| 2733708 | |
| CCOC(=O)CC(=O)C1=CC(F)=C(Cl)N=C1Cl |
| C10H8Cl2FNO3 | |
| MFCD00799535 | |
| ethyl 3-2,6-dichloro-5-fluoropyridin-3-yl-3-oxopropanoate, ethyl 3-2,6-dichloro-5-fluoro-3-pyridyl-3-oxopropionate, ethyl 2,6-dichloro-5-fluoronicotinoylacetate, ethyl 2,6-dichloro-5-fluoro pyridine-3-acetoacetate, ethyl 2,6-dichloro-5-fluoronicotinoyl acetate, ethyl 2,6-dichloro-5-fluornicotinoylacetate, ethyl 3-2,6-dichloro-5-fluoro-3-pyridiyl-3-oxopropanoate, 2,6-dichloro-5-fluoronicotinoyl acetic acid ethyl ester, ethyl 2,6-dichloro-5-fluoro nicotinylacetate, ethyl 3-2,6-dichloro-5-fluoro 3-pyridyl-3-oxopropanoate | |
| ethyl 3-(2,6-dichloro-5-fluoropyridin-3-yl)-3-oxopropanoate |
Safety and Handling
TSCA : No