accessibility menu, dialog, popup

UserName

H3COOOFClNCl

CAS RN 96568-04-6

IUPAC Name: ethyl 3-(2,6-dichloro-5-fluoropyridin-3-yl)-3-oxopropanoate
Synonyms: ethyl 3-(2,6-dichloro-5-fluoropyridin-3-yl)-3-oxopropanoate
Molecular Weight (g/mol): 280.08
Molecular Formula: C10H8Cl2FNO3
InChi Key: IEUHWNLWVMLHHC-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1=CC(F)=C(Cl)N=C1Cl
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)