missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. p-7031108
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3206503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3206503 Supplier Thermo Scientific Chemicals Supplier No. H3206503

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 306960-67-8
Molecular Formula C9H9F3N2O2
Molecular Weight (g/mol) 234.178
MDL Number MFCD07367386
InChI Key LYEKBQSEKSEXLT-UHFFFAOYSA-N
Synonym ethyl 4-methyl-2-trifluoromethyl pyrimidine-5-carboxylate, ethyl 2-trifluoromethyl-4-methyl-5-pyrimidine carboxylate, ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate, ethyl-2-trifluoromethyl-4-methyl-5-pyrimidine carboxylate, ethyl4-methyl-2-trifluoromethyl pyrimidine-5-carboxylate, 5-pyrimidinecarboxylicacid, 4-methyl-2-trifluoromethyl-, ethyl ester
PubChem CID 10609709
IUPAC Name ethyl 4-methyl-2-(trifluoromethyl)pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N=C1C)C(F)(F)F

Specifications

Boiling Point 54°C (0.5 mmHg)
Quantity 1 g
Formula Weight 234.18
Percent Purity 97%
Chemical Name or Material Ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only