missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Thiophenecarboxylate 98.0+%, TCI America™

Catalog No. p-7136488
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T090410ML 10 mL
T090425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T090410ML Supplier TCI America Supplier No. T090410ML

Chemical Identifiers

CAS 2810-04-0
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.20
MDL Number MFCD00005436
InChI Key JZGZKRJVTIRPOK-UHFFFAOYSA-N
Synonym ethyl 2-thiophenecarboxylate, ethyl 2-thenoate, ethyl-2-thiophenecarboxylate, thiophene-2-carboxylic acid ethyl ester, 2-thiophenecarboxylic acid ethyl ester, thiophene, 2-ethyl carboxylate, thiophenecarboxylic acid, ethyl ester, ethylthiophen-2-carboxylat, ethyl2-thiophenecarboxylate, acmc-209h0w
PubChem CID 76052
IUPAC Name ethyl thiophene-2-carboxylate
SMILES CCOC(=O)C1=CC=CS1

Specifications

Color Yellow
Boiling Point 218°C
Quantity 10 mL
Formula Weight 156.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Thiophenecarboxylate