missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Pyridylacetate 98.0+%, TCI America™

Catalog No. P10755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10755G 5 g
P107525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10755G Supplier TCI America Supplier No. P10755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2739-98-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00006359
InChI Key IUDKTVXSXWAKJO-UHFFFAOYSA-N
Synonym ethyl 2-pyridylacetate, ethyl 2-pyridin-2-yl acetate, ethyl 2-pyridinylacetate, ethyl pyridine-2-acetate, 2-pyridineacetic acid ethyl ester, ethyl 2-pyridineacetate, ethyl 2-2-pyridyl acetate, 2-pyridylacetic acid ethyl ester, ethyl pyridin-2-ylacetate, 2-pyridineacetic acid, ethyl ester
PubChem CID 75960
IUPAC Name ethyl 2-pyridin-2-ylacetate
SMILES CCOC(=O)CC1=CC=CC=N1

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 g
Formula Weight 165.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Pyridylacetate