accessibility menu, dialog, popup

UserName

CAS RN 2739-98-2

IUPAC Name: ethyl 2-(pyridin-2-yl)acetate
Synonyms: ethyl 2-(pyridin-2-yl)acetate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: IUDKTVXSXWAKJO-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=CC=N1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)