missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-pyridineacetate, 98%

Catalog No. p-7027321
Change view
Click to view available options
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AAB2398806 5 g
AAB2398814 25 g
AAB2398822 100 g
3 options

Catalog No. AAB2398806

Supplier: Thermo Scientific Chemicals B2398806

Only null left

CAS: 2739-98-2 | C9H11NO2 | 165.192 g/mol

Ethyl 2-pyridineacetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-pyridineacetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2739-98-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00006359
InChI Key IUDKTVXSXWAKJO-UHFFFAOYSA-N
Synonym ethyl 2-pyridylacetate, ethyl 2-pyridin-2-yl acetate, ethyl 2-pyridinylacetate, ethyl pyridine-2-acetate, 2-pyridineacetic acid ethyl ester, ethyl 2-pyridineacetate, ethyl 2-2-pyridyl acetate, 2-pyridylacetic acid ethyl ester, ethyl pyridin-2-ylacetate, 2-pyridineacetic acid, ethyl ester
PubChem CID 75960
IUPAC Name ethyl 2-pyridin-2-ylacetate
SMILES CCOC(=O)CC1=CC=CC=N1

Specifications

Density 1.084
Boiling Point 69°C to 70°C (0.05 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.497
Quantity 5 g
Beilstein 123817
Solubility Information Soluble in water.
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material Ethyl 2-pyridineacetate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-365-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only