missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Piperidinecarboxylate 98.0+%, TCI America™
Supplier: TCI America E083725G
Specifications
| Ethyl 2-Piperidinecarboxylate | |
| Yellow | |
| C8H16NO2 | |
| 25 g | |
| ethyl pipecolinate, ethyl 2-piperidinecarboxylate, ethyl pipecolate, pipecolinic acid ethyl ester, pipecolic acid ethyl ester, ethylpiperidine-2-carboxylate, 2-piperidinecarboxylic acid ethyl ester, 2-piperidinecarboxylic acid, ethyl ester, ethylpipecolinate, pubchem7720 | |
| CCOC(=O)[C@@H]1CCCC[NH2+]1 | |
| 158.22 | |
| 157.21 | |
| Liquid |
| 15862-72-3 | |
| 217°C | |
| MFCD00005980 | |
| 1993 | |
| SZIKRGHFZTYTIT-ZETCQYMHSA-O | |
| (2S)-2-(ethoxycarbonyl)piperidin-1-ium | |
| 27517 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 15862-72-3 | |
| 158.22 | |
| SZIKRGHFZTYTIT-ZETCQYMHSA-O | |
| 27517 | |
| CCOC(=O)[C@@H]1CCCC[NH2+]1 |
| C8H16NO2 | |
| MFCD00005980 | |
| ethyl pipecolinate, ethyl 2-piperidinecarboxylate, ethyl pipecolate, pipecolinic acid ethyl ester, pipecolic acid ethyl ester, ethylpiperidine-2-carboxylate, 2-piperidinecarboxylic acid ethyl ester, 2-piperidinecarboxylic acid, ethyl ester, ethylpipecolinate, pubchem7720 | |
| (2S)-2-(ethoxycarbonyl)piperidin-1-ium |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator