missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-pentynoate, 97+%

Catalog No. AAL0270306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0270306 5 g
AAL0270314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0270306 Supplier Thermo Scientific Chemicals Supplier No. L0270306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 55314-57-3 | C7H10O2 | 126.155 g/mol

Ethyl 2-pentynoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-pentynoate is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Store at 4°C. Store away from oxidizing agents. Keep away from heat. Incompatible with Oxidizing material. Acids

Chemical Identifiers

CAS 55314-57-3
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00015221
InChI Key XDPRPKSTFBPPHU-UHFFFAOYSA-N
PubChem CID 99169
IUPAC Name ethyl pent-2-ynoate
SMILES CCC#CC(=O)OCC

Specifications

Density 0.957
Boiling Point 178°C to 179°C
Flash Point 64°C (147°F)
Refractive Index 1.4395
Linear Formula CH3CH2C≡CCO2CH2CH3
Quantity 5 g
Beilstein 1750016
Synonym ethyl 2-pentynoate, 2-pentynoic acid ethyl ester, ethyl-2-pentynoate, ethyl pent-2-yn-1-oate, 2-pentynoic acid, ethyl ester, ethyl2-pentynoate, acmc-20al0p, 2-pentynoic acid, ethylester, pent-2-ynoic acid ethyl ester
Solubility Information Not miscible in water.
Formula Weight 126.16
Percent Purity 98%
Chemical Name or Material Ethyl 2-pentynoate
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 259-588-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only