CAS RN 55314-57-3
IUPAC Name:
ethyl pent-2-ynoate
Synonyms:
ethyl pent-2-ynoate
Molecular Weight (g/mol):
126.16
Molecular Formula:
C7H10O2
InChi Key:
XDPRPKSTFBPPHU-UHFFFAOYSA-N
SMILES:
CCOC(=O)C#CCC
1
–
1
of
1
results
1
Ethyl 2-pentynoate, 97+%
CAS: 55314-57-3 Molecular Formula: C7H10O2 Molecular Weight (g/mol): 126.155 MDL Number: MFCD00015221 InChI Key: XDPRPKSTFBPPHU-UHFFFAOYSA-N PubChem CID: 99169 IUPAC Name: ethyl pent-2-ynoate SMILES: CCC#CC(=O)OCC