Learn More
Ethyl 2-oximinooxamate, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 10489-74-4 |
|---|---|
| Molecular Formula | C4H8N2O3 |
| Molecular Weight (g/mol) | 132.12 |
| MDL Number | MFCD03013442 |
| InChI Key | QGYKRMZPOOILBA-UHFFFAOYSA-N |
| Synonym | ethyl 2-oximinooxamate, ethyl 2-amino-2-hydroxyimino acetate, ethyl z-2-amino-2-hydroxyimino acetate, ethyl 2-hydroxyamino-2-iminoacetate, ethyl n'-hydroxycarbamimidoyl formate, ethyl aminohydroxyiminoacetate, acetic acid, 2-amino-2-hydroxyimino-, ethyl ester, 2z, ethyl amino hydroxyimino ethanoate, ethyl n-hydroxycarbamimidoyl formate, amino-hydroxyimino-acetic acid ethyl ester |
| PubChem CID | 9614369 |
| IUPAC Name | ethyl (2Z)-2-amino-2-hydroxyiminoacetate |
| SMILES | CCOC(=O)C(=NO)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC426880050
|
Thermo Scientific Chemicals
426880050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 10489-74-4 | |
| 132.12 | |
| QGYKRMZPOOILBA-UHFFFAOYSA-N | |
| 9614369 | |
| CCOC(=O)C(=NO)N |
| C4H8N2O3 | |
| MFCD03013442 | |
| ethyl 2-oximinooxamate, ethyl 2-amino-2-hydroxyimino acetate, ethyl z-2-amino-2-hydroxyimino acetate, ethyl 2-hydroxyamino-2-iminoacetate, ethyl n'-hydroxycarbamimidoyl formate, ethyl aminohydroxyiminoacetate, acetic acid, 2-amino-2-hydroxyimino-, ethyl ester, 2z, ethyl amino hydroxyimino ethanoate, ethyl n-hydroxycarbamimidoyl formate, amino-hydroxyimino-acetic acid ethyl ester | |
| ethyl (2Z)-2-amino-2-hydroxyiminoacetate |
Specifications
| 10489-74-4 | |
| 97.5% min. (HPLC) | |
| C4H8N2O3 | |
| MFCD03013442 | |
| ethyl 2-oximinooxamate, ethyl 2-amino-2-hydroxyimino acetate, ethyl z-2-amino-2-hydroxyimino acetate, ethyl 2-hydroxyamino-2-iminoacetate, ethyl n'-hydroxycarbamimidoyl formate, ethyl aminohydroxyiminoacetate, acetic acid, 2-amino-2-hydroxyimino-, ethyl ester, 2z, ethyl amino hydroxyimino ethanoate, ethyl n-hydroxycarbamimidoyl formate, amino-hydroxyimino-acetic acid ethyl ester | |
| CCOC(=O)C(=NO)N | |
| 132.12 | |
| 132.12 | |
| Ethyl 2-oximinooxamate |
| Authentic | |
| Glass bottle | |
| HONHC(=NH)COOCH2CH3 | |
| 5g | |
| QGYKRMZPOOILBA-UHFFFAOYSA-N | |
| ethyl (2Z)-2-amino-2-hydroxyiminoacetate | |
| 9614369 | |
| 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Warning
RUO â Research Use Only