missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-nitropropionate, 96%

Catalog No. AAL1226103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1226103 1 g
AAL1226106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1226103 Supplier Thermo Scientific Chemicals Supplier No. L1226103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2531-80-8 | C5H9NO4 | 147.13 g/mol

Ethyl 2-nitropropionate is used in the preparation of (-)-trans-2,5-dimethylprolineethyl ester via enantioselective Michael reaction methyl vinyl ketone using modified dihydroquinine as catalyst, novel nitrogen-15-labeled nitrone spin trap, 5-ethoxycarbonyl-5-methyl-1-pyrroline N-oxide ([15N]EMPO) for detecting superoxide anion, 2-ethoxycarbonyl-2-nitropentanal. It is used for enantioselective Michael reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-nitropropionate is used in the preparation of (-)-trans-2,5-dimethylprolineethyl ester via enantioselective Michael reaction methyl vinyl ketone using modified dihydroquinine as catalyst, novel nitrogen-15-labeled nitrone spin trap, 5-ethoxycarbonyl-5-methyl-1-pyrroline N-oxide ([15N]EMPO) for detecting superoxide anion, 2-ethoxycarbonyl-2-nitropentanal. It is used for enantioselective Michael reactions.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.

Chemical Identifiers

CAS 2531-80-8
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00024818
InChI Key ZXBGJDZWJJFFQY-UHFFFAOYNA-N
Synonym ethyl 2-nitropropionate, propanoic acid, 2-nitro-, ethyl ester, propionic acid, 2-nitro-, ethyl ester, ethyl alpha-nitropropionate, acmc-1ce4b, ethyl .alpha.-nitropropionate, propanoicacid, 2-nitro-, ethyl ester
PubChem CID 97978
IUPAC Name ethyl 2-nitropropanoate
SMILES CCOC(=O)C(C)[N+]([O-])=O

Specifications

Density 1.13
Boiling Point 190°C to 195°C
Flash Point 89°C (192°F)
Refractive Index 1.421
Quantity 1 g
Beilstein 774734
Solubility Information Not miscible in water.
Formula Weight 147.13
Percent Purity 96%
Chemical Name or Material Ethyl 2-nitropropionate
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only