accessibility menu, dialog, popup

UserName

CAS RN 2531-80-8

IUPAC Name: ethyl 2-nitropropanoate
Synonyms: ethyl 2-nitropropanoate
Molecular Weight (g/mol): 147.13
Molecular Formula: C5H9NO4
InChi Key: ZXBGJDZWJJFFQY-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)[N+]([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)