missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-nitrobenzoylacetate, 97%

Catalog No. p-4089964
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC430040010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC430040010 Supplier Thermo Scientific Chemicals Supplier No. 430040010
Only null left

CAS: 52119-39-8 | C11H11NO5 | 237.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52119-39-8
Molecular Formula C11H11NO5
Molecular Weight (g/mol) 237.21
MDL Number MFCD03424821
InChI Key OWZNCVIBJQPNEF-UHFFFAOYSA-N
Synonym ethyl 3-2-nitrophenyl-3-oxopropanoate, ethyl 2-nitrobenzoylacetate, acmc-1awhb, ethyl 3-2-nitrophenyl-3-oxo-propanoate, ethyl o-nitrobenzoylacetate, ethyl 2-o-nitrobenzoyl acetate, ethyl 2-2-nitrobenzoyl acetate, ethyl 2-o-nitrobenzoyl-acetate, ethyl 2-nitrobenzoyl acetate
PubChem CID 10561837
IUPAC Name ethyl 3-(2-nitrophenyl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC=CC=C1[N+](=O)[O-]

Specifications

Melting Point 35.0°C to 36.0°C
Color Orange to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 237.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 2-nitrobenzoylacetate

RUO – Research Use Only