missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-nitrobenzoate, 97%

Catalog No. aaa1999422
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
Catalog No. Quantity
AAA1999422 100 g
AAA1999414 25 g
2 options

Catalog No. AAA1999422

Supplier: Thermo Scientific Chemicals A1999422

Only null left

CAS: 610-34-4 | C9H9NO4 | 195.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 610-34-4
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00014696
InChI Key CPNMAYYYYSWTIV-UHFFFAOYSA-N
Synonym ethyl o-nitrobenzoate, ethyl nitrobenzoate, benzoic acid, o-nitro-, ethyl ester, benzoic acid, 2-nitro-, ethyl ester, nitrobenzoesaureathylester, acmc-1aul6, 2-nitro-benzoic acid ethyl ester
PubChem CID 69123
IUPAC Name ethyl 2-nitrobenzoate
SMILES CCOC(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 26°C to 30°C
Boiling Point 172°C to 174°C (18 mmHg)
Quantity 100 g
Beilstein 2050729
Merck Index 14,3832
Formula Weight 195.18
Percent Purity 97%
Chemical Name or Material Ethyl 2-nitrobenzoate
EINECSNumber 210-220-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only