missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-morpholinobenzoate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC24323DA

Quantity 1g

 View more versions of this product

Catalog No. CC24323DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

192817-79-1
235.283
UIVDSGDHUXOYDW-UHFFFAOYSA-N
7148401
CCOC(=O)C1=CC=CC=C1N2CCOCC2
C13H17NO3
MFCD06204497
ethyl 2-morpholinobenzoate, ethyl 2-morpholin-4-yl benzoate, 2-morpholin-4-yl-benzoic acid ethyl ester, benzoic acid,2-4-morpholinyl-,ethyl ester, 2-morpholin-4-ylbenzoic acid ethyl ester, benzoic acid,2-4-morpholinyl-, ethyl ester
ethyl 2-morpholin-4-ylbenzoate

Specifications

Ethyl 2-morpholinobenzoate
96.5
97%
C13H17NO3
MFCD06204497
UIVDSGDHUXOYDW-UHFFFAOYSA-N
ethyl 2-morpholin-4-ylbenzoate
7148401
97%
192817-79-1
100.0
Amber glass bottle
1g
ethyl 2-morpholinobenzoate, ethyl 2-morpholin-4-yl benzoate, 2-morpholin-4-yl-benzoic acid ethyl ester, benzoic acid,2-4-morpholinyl-,ethyl ester, 2-morpholin-4-ylbenzoic acid ethyl ester, benzoic acid,2-4-morpholinyl-, ethyl ester
CCOC(=O)C1=CC=CC=C1N2CCOCC2
235.283
235.28
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents