missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methylpyridine-4-carboxylate hydrochloride, 95%

Catalog No. p-7038313
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5723903 1 g
AAH5723906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5723903 Supplier Thermo Scientific Chemicals Supplier No. H5723903
Only null left

CAS: 65550-32-5 | C9H12ClNO2 | 201.65 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.

Chemical Identifiers

CAS 65550-32-5
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD09991690
InChI Key DPRMHHCAVHGENN-UHFFFAOYSA-N
Synonym ethyl 2-methylisonicotinate hydrochloride, ethyl 2-methylpyridine-4-carboxylate hydrochloride, ethyl 2-methylisonicotinate hcl, 2-methylisonicotinic acid ethyl ester hydrochloride, acmc-20al0t, 2-methyl-isonicotinic acid ethyl ester hydrochloride, 2-methyl-4-pyridinecarboxylic acid ethyl ester hydrochloride, ethyl 2-methylpyridine-4-carboxylate-hydrogen chloride 1/1, 4-pyridinecarboxylic acid, 2-methyl-, ethyl ester, hydrochloride
PubChem CID 53229930
SMILES Cl.CCOC(=O)C1=CC(C)=NC=C1

Specifications

Melting Point 145°C to 151°C
Quantity 1 g
Sensitivity Moisture sensitive
IUPAC Name ethyl 2-methylpyridine-4-carboxylate;hydrochloride
Formula Weight 201.65
Percent Purity 95%
Chemical Name or Material Ethyl 2-methylpyridine-4-carboxylate hydrochloride
Hazard Category H318
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P280-P305+P351+P338-P310
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only