CAS RN 65550-32-5
IUPAC Name:
ethyl 2-methylpyridine-4-carboxylate hydrochloride
Synonyms:
ethyl 2-methylpyridine-4-carboxylate hydrochloride
Molecular Weight (g/mol):
201.65
Molecular Formula:
C9H12ClNO2
InChi Key:
DPRMHHCAVHGENN-UHFFFAOYSA-N
SMILES:
Cl.CCOC(=O)C1=CC(C)=NC=C1
1
–
1
of
1
results
1
Promotions Available
Ethyl 2-methylpyridine-4-carboxylate hydrochloride, 95%
Ethyl 2-methylpyridine-4-carboxylate hydrochloride, 95%
CAS: 65550-32-5 Molecular Formula: C9H12ClNO2 Molecular Weight (g/mol): 201.65 MDL Number: MFCD09991690 InChI Key: DPRMHHCAVHGENN-UHFFFAOYSA-N Synonym: ethyl 2-methylisonicotinate hydrochloride,ethyl 2-methylpyridine-4-carboxylate hydrochloride,ethyl 2-methylisonicotinate hcl,2-methylisonicotinic acid ethyl ester hydrochloride,acmc-20al0t,2-methyl-isonicotinic acid ethyl ester hydrochloride,2-methyl-4-pyridinecarboxylic acid ethyl ester hydrochloride,ethyl 2-methylpyridine-4-carboxylate-hydrogen chloride 1/1,4-pyridinecarboxylic acid, 2-methyl-, ethyl ester, hydrochloride PubChem CID: 53229930 SMILES: Cl.CCOC(=O)C1=CC(C)=NC=C1