missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Methylacetoacetate 95.0+%, TCI America™
Supplier: TCI America M0698250ML
Specifications
| Ethyl 2-Methylacetoacetate | |
| Yellow | |
| C7H12O3 | |
| 250 mL | |
| FNENWZWNOPCZGK-UHFFFAOYSA-N | |
| ethyl 2-methyl-3-oxobutanoate | |
| 701 | |
| ≥95.0% (GC) |
| 609-14-3 | |
| 185°C | |
| MFCD00009164 | |
| ethyl 2-methylacetoacetate, ethyl 2-acetylpropionate, ethyl-2-methyl acetoacetate, ethyl methylacetoacetate, ethyl 2-methyl-3-oxobutyrate, ethyl2-methylacetoacetate, butanoic acid, 2-methyl-3-oxo-, ethyl ester, 2-methylacetoacetic acid ethyl ester, ethyl alpha-methylacetylacetate, 2-methyl-3-oxobutanoic acid ethyl ester | |
| CCOC(=O)C(C)C(=O)C | |
| 144.17 | |
| 144.17 | |
| Liquid |
Chemical Identifiers
| 609-14-3 | |
| 144.17 | |
| FNENWZWNOPCZGK-UHFFFAOYSA-N | |
| ethyl 2-methyl-3-oxobutanoate |
| C7H12O3 | |
| MFCD00009164 | |
| 701 | |
| CCOC(=O)C(C)C(=O)C |
Safety and Handling
EINECSNumber : (2)-1505
TSCA : Yes