missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Methylacetoacetate 95.0+%, TCI America™

Catalog No. p-7132306
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M069825ML 25 mL
M0698250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M069825ML Supplier TCI America Supplier No. M069825ML
Only null left

Chemical Identifiers

CAS 609-14-3
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009164
InChI Key FNENWZWNOPCZGK-UHFFFAOYSA-N
PubChem CID 701
IUPAC Name ethyl 2-methyl-3-oxobutanoate
SMILES CCOC(=O)C(C)C(=O)C

Specifications

Color Yellow
Boiling Point 185°C
Quantity 25 mL
Synonym ethyl 2-methylacetoacetate, ethyl 2-acetylpropionate, ethyl-2-methyl acetoacetate, ethyl methylacetoacetate, ethyl 2-methyl-3-oxobutyrate, ethyl2-methylacetoacetate, butanoic acid, 2-methyl-3-oxo-, ethyl ester, 2-methylacetoacetic acid ethyl ester, ethyl alpha-methylacetylacetate, 2-methyl-3-oxobutanoic acid ethyl ester
Formula Weight 144.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Methylacetoacetate
EINECSNumber (2)-1505
TSCA Yes