missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methyl-4-pentenoate, 98%

CAS: 53399-81-8 | C8H14O2 | 142.20 g/mol

$49.78 - $368.95

Chemical Identifiers

CAS 53399-81-8
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00010235
InChI Key BDBGKYIBDXAVMX-UHFFFAOYNA-N
Synonym ethyl 2-methyl-4-pentenoate, 2-methyl-4-pentenoic acid ethyl ester, ethyl 2-methyl 4-pentenoate, 4-pentenoic acid, 2-methyl-, ethyl ester, ethyl 2-methylpent-4-en-1-oate, ethyl 2-methyl-4-pentanoate, fema no. 3489, pineapple pentenoate, acmc-1ao14, ethyl-2-methyl-4-pentenoate
PubChem CID 62024
IUPAC Name ethyl 2-methylpent-4-enoate
SMILES CCOC(=O)C(C)CC=C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1952606
View Documents
Thermo Scientific Chemicals
A1952606
5 g
Each for $49.78
Only null left
 
AAA1952614
View Documents
Thermo Scientific Chemicals
A1952614
25 g
Each for $128.15
Only null left
 
AAA1952622
View Documents
Thermo Scientific Chemicals
A1952622
100 g
Each for $368.95
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

53399-81-8
142.20
BDBGKYIBDXAVMX-UHFFFAOYNA-N
62024
CCOC(=O)C(C)CC=C
C8H14O2
MFCD00010235
ethyl 2-methyl-4-pentenoate, 2-methyl-4-pentenoic acid ethyl ester, ethyl 2-methyl 4-pentenoate, 4-pentenoic acid, 2-methyl-, ethyl ester, ethyl 2-methylpent-4-en-1-oate, ethyl 2-methyl-4-pentanoate, fema no. 3489, pineapple pentenoate, acmc-1ao14, ethyl-2-methyl-4-pentenoate
ethyl 2-methylpent-4-enoate

Specifications

53399-81-8
153°C to 155°C
C8H14O2
MFCD00010235
UN3272
ethyl 2-methyl-4-pentenoate, 2-methyl-4-pentenoic acid ethyl ester, ethyl 2-methyl 4-pentenoate, 4-pentenoic acid, 2-methyl-, ethyl ester, ethyl 2-methylpent-4-en-1-oate, ethyl 2-methyl-4-pentanoate, fema no. 3489, pineapple pentenoate, acmc-1ao14, ethyl-2-methyl-4-pentenoate
CCOC(=O)C(C)CC=C
142.20
142.2
Ethyl 2-methyl-4-pentenoate
0.88
40°C (104°F)
1.416
5 g
1753406
BDBGKYIBDXAVMX-UHFFFAOYNA-N
ethyl 2-methylpent-4-enoate
62024
98%
Videos
Safety and Handling

Safety and Handling

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 258-520-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only