missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (2-Methoxybenzoyl)acetate 98.0+%, TCI America™

Catalog No. E11201G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E11201G Supplier TCI America Supplier No. E11201G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41607-95-8
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD01311633
InChI Key KROPYAVVJDXRPH-UHFFFAOYSA-N
Synonym (2-Methoxybenzoyl)acetic Acid Ethyl Ester, Ethyl 3-(2-Methoxyphenyl)-3-oxopropionate, 3-(2-Methoxyphenyl)-3-oxopropionic Acid Ethyl Ester
PubChem CID 2759695
IUPAC Name ethyl 3-(2-methoxyphenyl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC=CC=C1OC

Specifications

Boiling Point 122°C
Quantity 1 g
Formula Weight 222.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (2-Methoxybenzoyl)acetate
EINECSNumber (3)-1745
TSCA No