accessibility menu, dialog, popup

UserName

H3COOOOH3C

CAS RN 41607-95-8

IUPAC Name: ethyl 3-(2-methoxyphenyl)-3-oxopropanoate
Synonyms: ethyl 3-(2-methoxyphenyl)-3-oxopropanoate
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: KROPYAVVJDXRPH-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1=CC=CC=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results