missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Methoxy-6-methylbenzoate 95.0+%, TCI America™

Catalog No. p-7132550
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
M11205G 5 g
M112025G 25 g
2 options

Catalog No. M11205G

Supplier: TCI America M11205G

Chemical Identifiers

CAS 6520-83-8
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00191666
InChI Key GQLSLEBNODXANJ-UHFFFAOYSA-N
Synonym 2-Methoxy-6-methylbenzoic Acid Ethyl Ester
PubChem CID 591060
IUPAC Name ethyl 2-methoxy-6-methylbenzoate
SMILES CCOC(=O)C1=C(C=CC=C1OC)C

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 g
Formula Weight 194.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Methoxy-6-methylbenzoate