missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-iodobenzoate, 98%

Catalog No. aal0765014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0765014 25 g
AAL0765006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0765014 Supplier Thermo Scientific Chemicals Supplier No. L0765014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1829-28-3 | C9H9IO2 | 276.07 g/mol

Ethyl 2-iodobenzoate is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-iodobenzoate is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

Solubility
Difficult to mix.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1829-28-3
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.07
MDL Number MFCD00051796
InChI Key QOUFDDUDXYJWHV-UHFFFAOYSA-N
Synonym 2-iodobenzoic acid ethyl ester, rarechem al bi 0562, ethyl iodobenzoate, ethyl-2-iodobenzoate, ethyl 2-iodanylbenzoate, acmc-1ag98, 2-ethoxycarbonyl iodobenzene, benzoic acid, 2-iodo-,ethyl ester, benzoic acid, 2-iodo-, ethyl ester
PubChem CID 2829347
IUPAC Name ethyl 2-iodobenzoate
SMILES CCOC(=O)C1=CC=CC=C1I

Specifications

Boiling Point 163°C to 165°C (23 mmHg)
Refractive Index 1.584
Quantity 25 g
Beilstein 2084297
Sensitivity Light sensitive
Solubility Information Difficult to mix.
Formula Weight 276.08
Percent Purity 98%
Chemical Name or Material Ethyl 2-iodobenzoate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only