missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Iodo-2-methylpropionate 94.0+%, TCI America™

Catalog No. p-7129389
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11995G 5 g
E119925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11995G Supplier TCI America Supplier No. E11995G

Chemical Identifiers

CAS 7425-55-0
Molecular Formula C6H11IO2
Molecular Weight (g/mol) 242.056
InChI Key AYVTVRNTNCEPSH-UHFFFAOYSA-N
Synonym 2-Iodo-2-methylpropionic Acid Ethyl Ester
PubChem CID 11299482
IUPAC Name ethyl 2-iodo-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)I

Specifications

Quantity 5 g
Formula Weight 242.06
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Iodo-2-methylpropionate
TSCA No
Recommended Storage Freezer