missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(Hydroxymethyl)acrylate (stabilized with HQ) 95.0+%, TCI America™

Catalog No. H09165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09165G 5 g
H091625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09165G Supplier TCI America Supplier No. H09165G
Only null left

Chemical Identifiers

CAS 10029-04-6
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD01673859
InChI Key SYGAXBISYRORDR-UHFFFAOYSA-N
Synonym 2-(Hydroxymethyl)acrylic Acid Ethyl Ester
PubChem CID 24827
IUPAC Name ethyl 2-(hydroxymethyl)prop-2-enoate
SMILES CCOC(=O)C(=C)CO

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 g
UN Number 2810
Formula Weight 130.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-(Hydroxymethyl)acrylate (stabilized with HQ)
EINECSNumber (2)-3861
RTECSNumber AT1835000
TSCA No
Recommended Storage Refrigerator