accessibility menu, dialog, popup

UserName

CAS RN 10029-04-6

H3COOCH2OH

CAS RN 10029-04-6

IUPAC Name: ethyl 2-(hydroxymethyl)prop-2-enoate
Synonyms: ethyl 2-(hydroxymethyl)prop-2-enoate
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: SYGAXBISYRORDR-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=C)CO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results