Learn More
Ethyl 2-hydroxyisobutyrate, 98%
CAS: 80-55-7 | C6H12O3 | 132.16 g/mol
$163.45 - $663.75
Chemical Identifiers
| CAS | 80-55-7 |
|---|---|
| Molecular Formula | C6H12O3 |
| Molecular Weight (g/mol) | 132.16 |
| MDL Number | MFCD00004458 |
| InChI Key | GFUIDHWFLMPAGY-UHFFFAOYSA-N |
| Synonym | ethyl 2-hydroxyisobutyrate, ethyl-2-hydroxyisobutyrate, ethyl 2-hydroxy-2-methylpropionate, ethyl 2-methyllactate, ethyl alpha-hydroxyisobutyrate, ethyl alpha-hydroxyisobutyrate, 2-methyllactic acid ethyl ester, a-hydroxyisobutyrate, a-hydroxyisobutyrate, unii-s1530mcv3x, unii-s1530mcv3x |
| PubChem CID | 6653 |
| IUPAC Name | ethyl 2-hydroxy-2-methylpropanoate |
| SMILES | CCOC(=O)C(C)(C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2359222
|
Thermo Scientific Chemicals
B2359222 |
100 g |
N/A
|
|
|||||
|
AAB2359236
|
Thermo Scientific Chemicals
B2359236 |
500 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 80-55-7 | |
| 132.16 | |
| GFUIDHWFLMPAGY-UHFFFAOYSA-N | |
| 6653 | |
| CCOC(=O)C(C)(C)O |
| C6H12O3 | |
| MFCD00004458 | |
| ethyl 2-hydroxyisobutyrate, ethyl-2-hydroxyisobutyrate, ethyl 2-hydroxy-2-methylpropionate, ethyl 2-methyllactate, ethyl alpha-hydroxyisobutyrate, ethyl alpha-hydroxyisobutyrate, 2-methyllactic acid ethyl ester, a-hydroxyisobutyrate, a-hydroxyisobutyrate, unii-s1530mcv3x, unii-s1530mcv3x | |
| ethyl 2-hydroxy-2-methylpropanoate |
Specifications
| 80-55-7 | |
| 149°C to 150°C | |
| C6H12O3 | |
| MFCD00004458 | |
| UN3272 | |
| ethyl 2-hydroxyisobutyrate, ethyl-2-hydroxyisobutyrate, ethyl 2-methyllactate, propanoic acid, 2-hydroxy-2-methyl-, ethyl ester, ethyl alpha-hydroxyisobutyrate, ethyl alpha-hydroxyisobutyrate, ethyl, ethyl, lactic acid, 2-methyl-, ethyl ester, lactic acid, 2-methyl-, ethyl ester, unii-s1530mcv3x | |
| CCOC(=O)C(C)(C)O | |
| 132.16 | |
| 132.16 | |
| Ethyl 2-hydroxyisobutyrate |
| 0.98 | |
| 44°C (111°F) | |
| 1.408 | |
| 100 g | |
| 878308 | |
| GFUIDHWFLMPAGY-UHFFFAOYSA-N | |
| ethyl 2-hydroxy-2-methylpropanoate | |
| 6653 | |
| 98% |
Safety and Handling
GHS H Statement
H226-H319-H335
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 201-290-3
RTECSNumber : OD5672000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only