missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Hydroxy-4-(trimethylsilyl)-3-butynoate 97.0+%, TCI America™

Catalog No. p-7130790
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08951G 1 g
H08955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08951G Supplier TCI America Supplier No. H08951G

Chemical Identifiers

CAS 66697-09-4
Molecular Formula C9H16O3Si
Molecular Weight (g/mol) 200.309
MDL Number MFCD00671544
InChI Key KGHJOCNNONUDLT-UHFFFAOYSA-N
Synonym 2-Hydroxy-4-(trimethylsilyl)-3-butynoic Acid Ethyl Ester
PubChem CID 44630343
IUPAC Name ethyl 2-hydroxy-4-trimethylsilylbut-3-ynoate
SMILES CCOC(=O)C(C#C[Si](C)(C)C)O

Specifications

Quantity 1 g
Formula Weight 200.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Hydroxy-4-(trimethylsilyl)-3-butynoate
TSCA No
Recommended Storage Refrigerator