accessibility menu, dialog, popup

UserName

H3COOSiCH3CH3H3COH

CAS RN 66697-09-4

IUPAC Name: ethyl 2-hydroxy-4-(trimethylsilyl)but-3-ynoate
Synonyms: ethyl 2-hydroxy-4-(trimethylsilyl)but-3-ynoate
Molecular Weight (g/mol): 200.31
Molecular Formula: C9H16O3Si
InChi Key: KGHJOCNNONUDLT-UHFFFAOYNA-N
SMILES: CCOC(=O)C(O)C#C[Si](C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results