missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Formylpropionate 97.0+%, TCI America™
$533.13 - $1496.84
Chemical Identifiers
| CAS | 27772-62-9 |
|---|---|
| Molecular Formula | C6H10O3 |
| Molecular Weight (g/mol) | 130.143 |
| MDL Number | MFCD00044012 |
| InChI Key | VVCYNVCCODBCOE-UHFFFAOYSA-N |
| Synonym | 2-Formylpropionic Acid Ethyl Ester |
| PubChem CID | 10898741 |
| IUPAC Name | ethyl 2-methyl-3-oxopropanoate |
| SMILES | CCOC(=O)C(C)C=O |
Chemical Identifiers
| 27772-62-9 | |
| 130.143 | |
| VVCYNVCCODBCOE-UHFFFAOYSA-N | |
| 10898741 | |
| CCOC(=O)C(C)C=O |
| C6H10O3 | |
| MFCD00044012 | |
| 2-Formylpropionic Acid Ethyl Ester | |
| ethyl 2-methyl-3-oxopropanoate |
Specifications
| 27772-62-9 | |
| C6H10O3 | |
| 1 g | |
| 2-Formylpropionic Acid Ethyl Ester | |
| CCOC(=O)C(C)C=O | |
| 130.143 | |
| 130.14 | |
| Liquid |
| Yellow | |
| MFCD00044012 | |
| 1993 | |
| VVCYNVCCODBCOE-UHFFFAOYSA-N | |
| ethyl 2-methyl-3-oxopropanoate | |
| 10898741 | |
| ≥97.0% (T) | |
| Ethyl 2-Formylpropionate |
Safety and Handling
TSCA : No
Recommended Storage : Freezer