missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Formylpropionate 97.0+%, TCI America™

Supplier:  TCI America F04115G

 View more versions of this product

Catalog No. F04115G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 2-Formylpropionate
Yellow
MFCD00044012
1993
VVCYNVCCODBCOE-UHFFFAOYSA-N
ethyl 2-methyl-3-oxopropanoate
10898741
≥97.0% (T)
27772-62-9
C6H10O3
5 g
2-Formylpropionic Acid Ethyl Ester
CCOC(=O)C(C)C=O
130.143
130.14
Liquid

Chemical Identifiers

27772-62-9
130.143
VVCYNVCCODBCOE-UHFFFAOYSA-N
10898741
CCOC(=O)C(C)C=O
C6H10O3
MFCD00044012
2-Formylpropionic Acid Ethyl Ester
ethyl 2-methyl-3-oxopropanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions