missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-fluoroisobutyrate, 97%, Thermo Scientific Chemicals

Catalog No. p-7031751
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3293106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3293106 Supplier Thermo Scientific Chemicals Supplier No. H3293106

Ethyl 2-fluoroisobutyrate is used as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-fluoroisobutyrate is used as an active pharmaceutical ingredient.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 55816-69-8
Molecular Formula C6H11FO2
Molecular Weight (g/mol) 134.15
MDL Number MFCD08062432
InChI Key CJRQQJKWNULSFQ-UHFFFAOYSA-N
Synonym ethyl 2-fluoroisobutyrate, propanoic acid, 2-fluoro-2-methyl-, ethyl ester, ethyl 2-fluoro-isobutyrate, ethyl 2-fluoro-2-methylpropionate, 2-fluoro-2-methylpropionic acid ethyl ester
PubChem CID 10898763
IUPAC Name ethyl 2-fluoro-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)F

Specifications

Boiling Point 122°C to 126°C
Quantity 5 g
UN Number UN1992
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 134.15
Percent Purity 97%
Chemical Name or Material Ethyl 2-fluoroisobutyrate
Hazard Category H226-H301+H311+H331
Hazard Statement GHS H Statement
H301-H311-H331-H226
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only