accessibility menu, dialog, popup

UserName

CAS RN 55816-69-8

IUPAC Name: ethyl 2-fluoro-2-methylpropanoate
Synonyms: ethyl 2-fluoro-2-methylpropanoate
Molecular Weight (g/mol): 134.15
Molecular Formula: C6H11FO2
InChi Key: CJRQQJKWNULSFQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)(C)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results