missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (2-Fluorobenzoyl)acetate 98.0+%, TCI America™
$219.48 - $766.21
Chemical Identifiers
| CAS | 1479-24-9 |
|---|---|
| Molecular Formula | C11H11FO3 |
| Molecular Weight (g/mol) | 210.204 |
| MDL Number | MFCD00000321 |
| InChI Key | YMUNUVJSNWUWDA-UHFFFAOYSA-N |
| Synonym | ethyl 3-2-fluorophenyl-3-oxopropanoate, ethyl 2-fluorobenzoyl acetate, ethyl 3-2-fluorophenyl-3-oxo-propionate, 3-2-fluoro-phenyl-3-oxo-propionic acid ethyl ester, ethyl 3-2-fluorophenyl-3-oxopropionate, 3-2-fluorophenyl-3-oxopropionic acid ethyl ester, benzenepropanoic acid, 2-fluoro-beta-oxo-, ethyl ester, acmc-1c1es, ethyl 2-fluorobenzoylacetate, ethyl-2-fluorobenzoylacetate |
| PubChem CID | 296342 |
| IUPAC Name | ethyl 3-(2-fluorophenyl)-3-oxopropanoate |
| SMILES | CCOC(=O)CC(=O)C1=CC=CC=C1F |
Chemical Identifiers
| 1479-24-9 | |
| 210.204 | |
| YMUNUVJSNWUWDA-UHFFFAOYSA-N | |
| 296342 | |
| CCOC(=O)CC(=O)C1=CC=CC=C1F |
| C11H11FO3 | |
| MFCD00000321 | |
| ethyl 3-2-fluorophenyl-3-oxopropanoate, ethyl 2-fluorobenzoyl acetate, ethyl 3-2-fluorophenyl-3-oxo-propionate, 3-2-fluoro-phenyl-3-oxo-propionic acid ethyl ester, ethyl 3-2-fluorophenyl-3-oxopropionate, 3-2-fluorophenyl-3-oxopropionic acid ethyl ester, benzenepropanoic acid, 2-fluoro-beta-oxo-, ethyl ester, acmc-1c1es, ethyl 2-fluorobenzoylacetate, ethyl-2-fluorobenzoylacetate | |
| ethyl 3-(2-fluorophenyl)-3-oxopropanoate |
Specifications
| 1479-24-9 | |
| 108°C | |
| MFCD00000321 | |
| ethyl 3-2-fluorophenyl-3-oxopropanoate, ethyl 2-fluorobenzoyl acetate, ethyl 3-2-fluorophenyl-3-oxo-propionate, 3-2-fluoro-phenyl-3-oxo-propionic acid ethyl ester, ethyl 3-2-fluorophenyl-3-oxopropionate, 3-2-fluorophenyl-3-oxopropionic acid ethyl ester, benzenepropanoic acid, 2-fluoro-beta-oxo-, ethyl ester, acmc-1c1es, ethyl 2-fluorobenzoylacetate, ethyl-2-fluorobenzoylacetate | |
| CCOC(=O)CC(=O)C1=CC=CC=C1F | |
| 210.204 | |
| 210.20 | |
| Liquid |
| Yellow | |
| C11H11FO3 | |
| 5 g | |
| YMUNUVJSNWUWDA-UHFFFAOYSA-N | |
| ethyl 3-(2-fluorophenyl)-3-oxopropanoate | |
| 296342 | |
| ≥98.0% (GC) | |
| Ethyl (2-Fluorobenzoyl)acetate |
Safety and Handling
TSCA : No