missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Cyanopropionate 97.0+%, TCI America™
$89.48 - $274.02
Chemical Identifiers
| CAS | 1572-99-2 |
|---|---|
| Molecular Formula | C6H9NO2 |
| Molecular Weight (g/mol) | 127.143 |
| MDL Number | MFCD00013812 |
| InChI Key | MIHRVXYXORIINI-UHFFFAOYSA-N |
| Synonym | 2-Cyanopropionic Acid Ethyl Ester |
| PubChem CID | 98018 |
| ChEBI | CHEBI:51925 |
| IUPAC Name | ethyl 2-cyanopropanoate |
| SMILES | CCOC(=O)C(C)C#N |
Chemical Identifiers
| 1572-99-2 | |
| 127.143 | |
| MIHRVXYXORIINI-UHFFFAOYSA-N | |
| 98018 | |
| ethyl 2-cyanopropanoate |
| C6H9NO2 | |
| MFCD00013812 | |
| 2-Cyanopropionic Acid Ethyl Ester | |
| CHEBI:51925 | |
| CCOC(=O)C(C)C#N |
Specifications
| 1572-99-2 | |
| 193°C | |
| MFCD00013812 | |
| 3276 | |
| MIHRVXYXORIINI-UHFFFAOYSA-N | |
| ethyl 2-cyanopropanoate | |
| 98018 | |
| 127.14 | |
| Liquid |
| Yellow | |
| C6H9NO2 | |
| 5 mL | |
| 2-Cyanopropionic Acid Ethyl Ester | |
| CCOC(=O)C(C)C#N | |
| 127.143 | |
| CHEBI:51925 | |
| ≥97.0% (GC) | |
| Ethyl 2-Cyanopropionate |
Safety and Handling
TSCA : No