accessibility menu, dialog, popup

UserName

CAS RN 1572-99-2

H3COOCH3N

CAS RN 1572-99-2

IUPAC Name: ethyl 2-cyano-2-methylacetate
Synonyms: ethyl 2-cyanopropionate
Molecular Weight (g/mol): 127.14
Molecular Formula: C6H9NO2
InChi Key: MIHRVXYXORIINI-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results