missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-cyanoacetoacetate

Catalog No. p-7059022
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4379204 2 g
AA4379209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4379204 Supplier Thermo Scientific Chemicals Supplier No. 04379204

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 634-55-9 | C7H9NO3 | 155.15 g/mol

It is used as a pharmaceutical intermediate. It is also used in the synthesis of fluorine-containing alkyl 2-cyano-3-oxocarboxylates and also in the syntheses of pyrazolo [1, 2-a] pyrazole and pyrazolo [5, 1-b] [1, 3] oxazine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 634-55-9
Molecular Formula C7H9NO3
Molecular Weight (g/mol) 155.15
MDL Number MFCD00043487
InChI Key NWOKVFOTWMZMHL-UHFFFAOYNA-N
Synonym ethyl 2-cyanoacetoacetate, ethyl 2-acetylcyanoacetate, ethyl a-cyanoacetoacetate, ethyl alpha-cyanoacetoacetate, butanoic acid, 2-cyano-3-oxo-, ethyl ester, ethyl2-acetylcyanoacetate, ethyl 2-cyano-3-oxo-butanoate, 2-cyanoacetoacetic acid ethyl ester, 2-cyano-3-oxo-butyric acid ethyl ester, zlchem 939
PubChem CID 136455
ChEBI CHEBI:51924
IUPAC Name ethyl 2-cyano-3-oxobutanoate
SMILES CCOC(=O)C(C#N)C(C)=O

Specifications

Quantity 2 g
UN Number UN3276
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 155.16
Physical Form Liquid
Chemical Name or Material Ethyl 2-cyanoacetoacetate
Hazard Category H301+H311-H330
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only