accessibility menu, dialog, popup

UserName

CAS RN 634-55-9

IUPAC Name: ethyl 2-cyano-3-oxobutanoate
Synonyms: ethyl 2-cyanoacetoacetate
Molecular Weight (g/mol): 155.15
Molecular Formula: C7H9NO3
InChi Key: NWOKVFOTWMZMHL-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C#N)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results