missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Chloropropionate 97.0+%, TCI America™

Catalog No. p-7124610
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C079025ML 25 mL
C0790100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C079025ML Supplier TCI America Supplier No. C079025ML
Only null left

Chemical Identifiers

CAS 535-13-7
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00000864
InChI Key JEAVBVKAYUCPAQ-UHFFFAOYNA-N
PubChem CID 10807
IUPAC Name ethyl 2-chloropropanoate
SMILES CCOC(=O)C(C)Cl

Specifications

Color Yellow
Boiling Point 147°C
Quantity 25 mL
UN Number 2935
Synonym ethyl 2-chloropropionate, 2-chloropropionic acid ethyl ester, propanoic acid, 2-chloro-, ethyl ester, ethyl-2-chloropropionate, 2-chloropropionic acid, ethyl ester, ethyl alpha-chloropropionate, ethyl .alpha.-chloropropionate, propionic acid, 2-chloro-, ethyl ester, +-ethyl 2-chloropropionate, ethyl ?-chloropropionate
Formula Weight 136.58
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Chloropropionate
EINECSNumber (2)-3301
TSCA Yes