CAS RN 535-13-7
IUPAC Name:
ethyl 2-chloropropanoate
Synonyms:
ethyl α-chloropropionate
Molecular Weight (g/mol):
136.58
Molecular Formula:
C5H9ClO2
InChi Key:
JEAVBVKAYUCPAQ-UHFFFAOYNA-N
SMILES:
CCOC(=O)C(C)Cl
1
–
1
of
1
results
1
Ethyl 2-Chloropropionate 97.0+%, TCI America™
CAS: 535-13-7 Molecular Formula: C5H9ClO2 Molecular Weight (g/mol): 136.58 MDL Number: MFCD00000864 InChI Key: JEAVBVKAYUCPAQ-UHFFFAOYNA-N PubChem CID: 10807 IUPAC Name: ethyl 2-chloropropanoate SMILES: CCOC(=O)C(C)Cl