missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloronicotinate, 98%

CAS: 1452-94-4 | C8H8ClNO2 | 185.607 g/mol

Supplier:  Thermo Scientific Chemicals B2035918

 View more versions of this product

Catalog No. AAB2035918


Only null left
Explore available promotions

Description

Description

Ethyl 2-chloronicotinate,is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-chloronicotinate,is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

Ethyl 2-chloronicotinate
C8H8ClNO2
50 g
PMIMPBYTPPRBGD-UHFFFAOYSA-N
ethyl 2-chloropyridine-3-carboxylate
2796614
98%
1452-94-4
MFCD00674129
ethyl 2-chloronicotinate, 2-chloronicotinic acid ethyl ester, ethyl-2-chloronicotinate, 2-chloro-nicotinic acid ethyl ester, ethyl 2-chloro-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 2-chloro-, ethyl ester, 2-chloro-3-pyridinecarboxylic acid ethyl ester, ethyl 2chloronicotinate, acmc-1brbe, ethyl 2-chloro-nicotinate
CCOC(=O)C1=C(N=CC=C1)Cl
185.607
185.61

Chemical Identifiers

1452-94-4
185.607
PMIMPBYTPPRBGD-UHFFFAOYSA-N
2796614
CCOC(=O)C1=C(N=CC=C1)Cl
C8H8ClNO2
MFCD00674129
ethyl 2-chloronicotinate, 2-chloronicotinic acid ethyl ester, ethyl-2-chloronicotinate, 2-chloro-nicotinic acid ethyl ester, ethyl 2-chloro-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 2-chloro-, ethyl ester, 2-chloro-3-pyridinecarboxylic acid ethyl ester, ethyl 2chloronicotinate, acmc-1brbe, ethyl 2-chloro-nicotinate
ethyl 2-chloropyridine-3-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only