missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloronicotinate, 98%

Catalog No. p-7025047
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2035909 10 g
AAB2035918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2035909 Supplier Thermo Scientific Chemicals Supplier No. B2035909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1452-94-4 | C8H8ClNO2 | 185.607 g/mol

Ethyl 2-chloronicotinate,is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-chloronicotinate,is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 1452-94-4
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
MDL Number MFCD00674129
InChI Key PMIMPBYTPPRBGD-UHFFFAOYSA-N
Synonym ethyl 2-chloronicotinate, 2-chloronicotinic acid ethyl ester, ethyl-2-chloronicotinate, 2-chloro-nicotinic acid ethyl ester, ethyl 2-chloro-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 2-chloro-, ethyl ester, 2-chloro-3-pyridinecarboxylic acid ethyl ester, ethyl 2chloronicotinate, acmc-1brbe, ethyl 2-chloro-nicotinate
PubChem CID 2796614
IUPAC Name ethyl 2-chloropyridine-3-carboxylate
SMILES CCOC(=O)C1=C(N=CC=C1)Cl

Specifications

Quantity 10 g
Formula Weight 185.61
Percent Purity 98%
Chemical Name or Material Ethyl 2-chloronicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only