Learn More
Ethyl 2-chloronicotinate, 98%
CAS: 1452-94-4 | C8H8ClNO2 | 185.607 g/mol
Supplier: Thermo Scientific Chemicals B2035909
Description
Ethyl 2-chloronicotinate,is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 2-chloronicotinate,is used as a pharmaceutical intermediate.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
| Ethyl 2-chloronicotinate | |
| C8H8ClNO2 | |
| 10 g | |
| PMIMPBYTPPRBGD-UHFFFAOYSA-N | |
| ethyl 2-chloropyridine-3-carboxylate | |
| 2796614 | |
| 98% |
| 1452-94-4 | |
| MFCD00674129 | |
| ethyl 2-chloronicotinate, 2-chloronicotinic acid ethyl ester, ethyl-2-chloronicotinate, 2-chloro-nicotinic acid ethyl ester, ethyl 2-chloro-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 2-chloro-, ethyl ester, 2-chloro-3-pyridinecarboxylic acid ethyl ester, ethyl 2chloronicotinate, acmc-1brbe, ethyl 2-chloro-nicotinate | |
| CCOC(=O)C1=C(N=CC=C1)Cl | |
| 185.607 | |
| 185.61 |
Chemical Identifiers
| 1452-94-4 | |
| 185.607 | |
| PMIMPBYTPPRBGD-UHFFFAOYSA-N | |
| 2796614 | |
| CCOC(=O)C1=C(N=CC=C1)Cl |
| C8H8ClNO2 | |
| MFCD00674129 | |
| ethyl 2-chloronicotinate, 2-chloronicotinic acid ethyl ester, ethyl-2-chloronicotinate, 2-chloro-nicotinic acid ethyl ester, ethyl 2-chloro-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 2-chloro-, ethyl ester, 2-chloro-3-pyridinecarboxylic acid ethyl ester, ethyl 2chloronicotinate, acmc-1brbe, ethyl 2-chloro-nicotinate | |
| ethyl 2-chloropyridine-3-carboxylate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only