missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Chlorobenzoate 98.0+%, TCI America™

Catalog No. C167125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C167125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C167125G Supplier TCI America Supplier No. C167125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 7335-25-3
Color Colorless
Boiling Point 243°C
Molecular Formula C9H9ClO2
Quantity 25 g
MDL Number MFCD00016338
Synonym 2-chlorobenzoic acid ethyl ester, ethyl o-chlorobenzoate, benzoic acid, 2-chloro-, ethyl ester, ethyl-2-chlorobenzoate, ethyl2-chlorobenzoate, pubchem3703, acmc-1bfxj, ethyl 2-chloranylbenzoate, chlorobenzoic acid ethyl ester, ksc498a5r
InChI Key RETLCWPMLJPOTP-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CC=CC=C1Cl
IUPAC Name ethyl 2-chlorobenzoate
Molecular Weight (g/mol) 184.62
PubChem CID 81783
Formula Weight 184.62
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Chlorobenzoate
Show More Show Less